missing translation for 'onlineSavingsMsg'
Learn More

9,9-Di-n-octylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15491588
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491588 1 g 1g
15412076 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491588 Supplier Thermo Scientific Alfa Aesar Supplier No. H55373.03
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 258865-48-4 | Molecular Formula: C29H44B2O4 | Molecular Weight: 478.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 258865-48-4
Molecular Formula C29H44B2O4
Molecular Weight (g/mol) 478.29
MDL Number MFCD03701608
InChI Key HURJMQMZDPOUOU-UHFFFAOYSA-N
Synonym 9,9-dioctylfluorene-2,7-diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl diboronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyldiboronic acid, 7-borono-9,9-dioctylfluoren-2-yl boronic acid, boronic acid, 9,9-dioctyl-9h-fluorene-2,7-diyl bis, boronic acid,b,b'-9,9-dioctyl-9h-fluorene-2,7-diyl bis, acmc-209gn0, 7-borono-9,9-dioctyl-fluoren-2-yl boronic acid, 7-borono-9,9-dioctyl-2-fluorenyl boronic acid
PubChem CID 4283240
IUPAC Name (7-borono-9,9-dioctylfluoren-2-yl)boronic acid
SMILES CCCCCCCCC1(CCCCCCCC)C2=CC(=CC=C2C2=C1C=C(C=C2)B(O)O)B(O)O

Specifications

Quantity 1 g
Formula Weight 478.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 9,9-Di-n-octylfluorene-2,7-diboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.