accessibility menu, dialog, popup

UserName

CAS RN 25978-74-9

H3COOOCH3N

CAS RN 25978-74-9

IUPAC Name: methyl 3-cyano-4-methoxybenzoate
Synonyms: methyl 3-cyano-4-methoxybenzoate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: RYJSFYBJYKFNCF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(C#N)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results