missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-cyano-4-methoxybenzoate, 97%

Product Code. 11317566
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317566 5 g 5g
11307566 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317566 Supplier Thermo Scientific Alfa Aesar Supplier No. L11107.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25978-74-9 | C10H9NO3 | 191.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25978-74-9
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD00052930
InChI Key RYJSFYBJYKFNCF-UHFFFAOYSA-N
Synonym benzoic acid, 3-cyano-4-methoxy-, methyl ester, acmc-1cj4t, methyl 3-cyano-4-methoxy-benzoate, methyl 3-cyano-4-methoxybenzoate., methyl 3-cyano-4-methyloxy benzoate, 3-cyano-4-methoxy-benzoic acid methyl ester, benzoicacid,3-cyano-4-methoxy-,methyl ester
PubChem CID 2801011
IUPAC Name methyl 3-cyano-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1)C(=O)OC)C#N

Specifications

Melting Point 123°C to 127°C
Quantity 5 g
UN Number UN3439
Beilstein 2374769
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material Methyl 3-cyano-4-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.