accessibility menu, dialog, popup

UserName

CH3FHOCl

CAS RN 261762-83-8

IUPAC Name: (2-chloro-6-fluoro-3-methylphenyl)methanol
Synonyms: (2-chloro-6-fluoro-3-methylphenyl)methanol
Molecular Weight (g/mol): 174.6
Molecular Formula: C8H8ClFO
InChi Key: PQHOAHOMVOQUAU-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(CO)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results