accessibility menu, dialog, popup

UserName

FFFFBr

CAS RN 261951-84-2

IUPAC Name: 1-(bromomethyl)-3-fluoro-2-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-3-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: HTWOLWSZQAPKQA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CBr)=C1C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results