accessibility menu, dialog, popup

UserName

CH3OBrFFF

CAS RN 261951-89-7

IUPAC Name: 4-(bromomethyl)-1-methoxy-2-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-1-methoxy-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: FZICFAROPYFJMF-UHFFFAOYSA-N
SMILES: COC1=CC=C(CBr)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results