accessibility menu, dialog, popup

UserName

NSOOH

CAS RN 266369-49-7

IUPAC Name: 4-(1,3-thiazol-2-yl)benzoic acid
Synonyms: 4-(1,3-thiazol-2-yl)benzoic acid
Molecular Weight (g/mol): 205.23
Molecular Formula: C10H7NO2S
InChi Key: NCAPRAZCSUNRLM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results