missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Thiazol-2-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10613815
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10613815 250 mg Amber Glass Bottle 250mg
10326614 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10613815 Supplier Thermo Scientific Maybridge Supplier No. CC40201CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 266369-49-7
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
MDL Number MFCD08275675
InChI Key NCAPRAZCSUNRLM-UHFFFAOYSA-N
Synonym 4-1,3-thiazol-2-yl benzoic acid, 4-thiazol-2-yl benzoic acid, 4-2-thiazolyl benzoic acid, 4-thiazol-2-yl benzoicacid, benzoic acid,4-2-thiazolyl
PubChem CID 23510471
IUPAC Name 4-(1,3-thiazol-2-yl)benzoic acid
SMILES C1=CC(=CC=C1C2=NC=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Thiazol-2-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.