accessibility menu, dialog, popup

UserName

CAS RN 27684-84-0

BrHONOO

CAS RN 27684-84-0

IUPAC Name: 5-bromo-2-nitrophenol
Synonyms: 5-bromo-2-nitrophenol
Molecular Weight (g/mol): 218.01
Molecular Formula: C6H4BrNO3
InChi Key: DTWHNSNSUBKGTC-UHFFFAOYSA-N
SMILES: OC1=C(C=CC(Br)=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)