CAS RN 2810-04-0
IUPAC Name:
ethyl thiophene-2-carboxylate
Synonyms:
thiophene, 2-ethyl carboxylate
Molecular Weight (g/mol):
156.2
Molecular Formula:
C7H8O2S
InChi Key:
JZGZKRJVTIRPOK-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CS1
1
–
2
of
2
results
1
Ethyl thiophene-2-carboxylate, 98+%
CAS: 2810-04-0 Molecular Formula: C7H8O2S Molecular Weight (g/mol): 156.20 MDL Number: MFCD00005436 InChI Key: JZGZKRJVTIRPOK-UHFFFAOYSA-N PubChem CID: 76052 IUPAC Name: ethyl thiophene-2-carboxylate SMILES: CCOC(=O)C1=CC=CS1
2
Ethyl Thiophene-2-carboxylate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.