accessibility menu, dialog, popup

UserName

CAS RN 2835-95-2

IUPAC Name: 5-amino-2-methylphenol
Synonyms: 5-amino-o-cresol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: DBFYESDCPWWCHN-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)