missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylphenol, 97%, Thermo Scientific Chemicals

Product Code. 15422719
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422719 100 g 100g
15432719 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422719 Supplier Thermo Scientific Alfa Aesar Supplier No. H66252.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2835-95-2 | Molecular Formula: C7H9NO | Molecular Weight: 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-95-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00043922
InChI Key DBFYESDCPWWCHN-UHFFFAOYSA-N
Synonym 5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
PubChem CID 17818
IUPAC Name 5-amino-2-methylphenol
SMILES CC1=C(C=C(C=C1)N)O

Specifications

Melting Point 160°C to 162°C
Color Brown
Quantity 100 g
Beilstein 2802317
Formula Weight 123.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Amino-2-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.