accessibility menu, dialog, popup

UserName

CAS RN 28399-31-7

CH3OOH

CAS RN 28399-31-7

IUPAC Name: 8-{2-[(2-pentylcyclopropyl)methyl]cyclopropyl}octanoic acid
Synonyms: 8-{2-[(2-pentylcyclopropyl)methyl]cyclopropyl}octanoic acid
Molecular Weight (g/mol): 308.51
Molecular Formula: C20H36O2
InChi Key: CONYTTFKIUJZOF-UHFFFAOYNA-N
SMILES: CCCCCC1CC1CC1CC1CCCCCCCC(O)=O
Color 
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results