CAS RN 28448-85-3
CAS RN 28448-85-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bavachalcone, MedChemExpress
MedChemExpress Bavachalcone is a major bioactive compounds isolated from Psoralea corylifolia L.