accessibility menu, dialog, popup

UserName

FBrNH2HCl

CAS RN 289038-14-8

IUPAC Name: 1-(2-bromo-4-fluorophenyl)methanamine hydrochloride
Synonyms: 1-(2-bromo-4-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 240.5
Molecular Formula: C7H8BrClFN
InChi Key: PBCSNHSXUSMNOV-UHFFFAOYSA-N
SMILES: Cl.NCC1=C(Br)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results