CAS RN 2924-09-6
IUPAC Name:
5-bromo-2-fluoroaniline
Synonyms:
5-bromo-2-fluoroaniline
Molecular Weight (g/mol):
190.02
Molecular Formula:
C6H5BrFN
InChi Key:
ADWKOCXRCRSMLQ-UHFFFAOYSA-N
SMILES:
NC1=C(F)C=CC(Br)=C1
1
–
1
of
1
results
1
5-Bromo-2-fluoroaniline, 98%
CAS: 2924-09-6 Molecular Formula: C6H5BrFN Molecular Weight (g/mol): 190.02 MDL Number: MFCD03094181 InChI Key: ADWKOCXRCRSMLQ-UHFFFAOYSA-N