missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluoroaniline, 98%

Product Code. 15428248
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428248 100 g 100g
15418248 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428248 Supplier Thermo Scientific Alfa Aesar Supplier No. H61678.22
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2924-09-6 | C6H5BrFN | 190.02 g/mol

5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-fluoroaniline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2924-09-6
Molecular Formula C6H5BrFN
Molecular Weight (g/mol) 190.02
MDL Number MFCD03094181
InChI Key ADWKOCXRCRSMLQ-UHFFFAOYSA-N

Specifications

Melting Point 27°C
Boiling Point 105°C (12 mmHg)
Quantity 100 g
UN Number UN2811
Beilstein 1241528
Synonym 5-bromo-2-fluorobenzenamine, 2-fluoro-5-bromoaniline, 5-bromo-2-fluoro aniline, 5-bromo-2-fluorophenylamine, benzenamine, 5-bromo-2-fluoro, 5-bromo-2-fluoro-phenylamine, 1-amino-5-bromo-2-fluorobenzene, 5-bromo-2fluoroaniline, pubchem13482, acmc-1cjje
Solubility Information Slightly soluble in water.
SMILES C1=CC(=C(C=C1Br)N)F
IUPAC Name 5-bromo-2-fluoroaniline
PubChem CID 2769407
Formula Weight 190.01
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.