accessibility menu, dialog, popup

UserName

HClH2NNHF

CAS RN 2924-15-4

IUPAC Name: hydrogen (2-fluorophenyl)hydrazine chloride
Synonyms: hydrogen (2-fluorophenyl)hydrazine chloride
Molecular Weight (g/mol): 162.59
Molecular Formula: C6H8ClFN2
InChi Key: VFZYLSYYMHFPSY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results