missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylhydrazine hydrochloride, 98%

Product Code. 11414724
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414724 5 g 5g
11404724 1 g 1g
11424724 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414724 Supplier Thermo Scientific Alfa Aesar Supplier No. A13716.06
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2924-15-4 | C6H8ClFN2 | 162.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2924-15-4
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.592
MDL Number MFCD00012927
InChI Key VFZYLSYYMHFPSY-UHFFFAOYSA-N
Synonym 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m
PubChem CID 2723910
IUPAC Name (2-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)NN)F.Cl

Specifications

Melting Point ∼205°C (decomposition)
Quantity 5 g
Beilstein 3627985
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 2-Fluorophenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.