accessibility menu, dialog, popup

UserName

CH3NNHSS

CAS RN 29490-19-5

IUPAC Name: 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms: 5-methyl-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol): 132.2
Molecular Formula: C3H4N2S2
InChi Key: FPVUWZFFEGYCGB-UHFFFAOYSA-N
SMILES: CC1=NNC(=S)S1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results