accessibility menu, dialog, popup

UserName

NH2N

CAS RN 2973-50-4

IUPAC Name: 2-(2-aminophenyl)acetonitrile
Synonyms: 2-(2-aminophenyl)acetonitrile
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: LMDPYYUISNUGGT-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CC#N
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results