missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenylacetonitrile, 97%

Product Code. 15482709
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482709 5 g 5g
15472709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482709 Supplier Thermo Scientific Alfa Aesar Supplier No. H66243.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2973-50-4 | C8H8N2 | 132.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-50-4
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.166
MDL Number MFCD00091082
InChI Key LMDPYYUISNUGGT-UHFFFAOYSA-N
Synonym 2-2-aminophenyl acetonitrile, 2-aminophenylacetonitrile, 2-aminobenzyl cyanide, 2-aminophenyl acetonitrile, 2-aminobenzylcyanide, o-aminobenzyl cyanide, o-aminophenylacetonitrile, 2-aminobenzeneacetonitrile, 2-2-aminophenyl ethanenitrile, benzeneacetonitrile, 2-amino
PubChem CID 76307
IUPAC Name 2-(2-aminophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)N

Specifications

Melting Point 69°C to 72°C
Color White
Quantity 5 g
UN Number UN3439
Beilstein 2716483
Formula Weight 132.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Aminophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.