accessibility menu, dialog, popup

UserName

CAS RN 298-81-7

H3COOOO

CAS RN 298-81-7

IUPAC Name: 9-methoxy-7H-furo[3,2-g]chromen-7-one
Synonyms: methoxsalen 8 methoxypsoralen meladinina meladinine 8-methoxypsoralen methoxsalen (product) sanofi synthelabo brand of methoxsalen ultra mop ultramop methoxsalen canderm brand
Molecular Weight (g/mol): 216.19
Molecular Formula: C12H8O4
InChi Key: QXKHYNVANLEOEG-UHFFFAOYSA-N
SMILES: COC1=C2OC=CC2=CC2=C1OC(=O)C=C2
Percent Purity 
  • (5)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)