accessibility menu, dialog, popup

UserName

ClClCH3NNONOHNNHH

CAS RN 29868-97-1

IUPAC Name: dihydrogen 2-[2-(4-methylpiperazin-1-yl)acetyl]-2,4,9-triazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one dichloride
Synonyms: pirenzepine hydrochloride
Molecular Weight (g/mol): 424.33
Molecular Formula: C19H23Cl2N5O2
InChi Key: FFNMBRCFFADNAO-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN1CCN(CC(=O)N2C3=CC=CC=C3C(=O)NC3=CC=CN=C23)CC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)