missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine dihydrochloride, 99%

Product Code. 15444659
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444659 100 mg 100mg
15424659 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15444659 Supplier Thermo Scientific Alfa Aesar Supplier No. J62252.MC
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A M1 muscarinic receptor antagonist | CAS: 29868-97-1 | C19H23Cl2N5O2 | 424.326 g/mol

A M1 muscarinic receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29868-97-1
Molecular Formula C19H23Cl2N5O2
Molecular Weight (g/mol) 424.326
MDL Number MFCD00055214
InChI Key FFNMBRCFFADNAO-UHFFFAOYSA-N
Synonym pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
PubChem CID 71405
ChEBI CHEBI:32014
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
SMILES CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

Specifications

Melting Point 192°C to 198°C
Color White
Quantity 100 mg
Sensitivity Hygroscopic
Merck Index 14,7491
Formula Weight 424.32
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Pirenzepine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.