accessibility menu, dialog, popup

UserName

CAS RN 29915-38-6

HOOHHOHNSOHOO

CAS RN 29915-38-6

IUPAC Name: 3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propane-1-sulfonic acid
Synonyms: taps (buffer)
Molecular Weight (g/mol): 243.27
Molecular Formula: C7H17NO6S
InChi Key: YNLCVAQJIKOXER-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)NCCCS(O)(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (3)
Quantity 
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (5)
  • (1)
pH 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)