accessibility menu, dialog, popup

UserName

H3COSiOH3COCH3

CAS RN 2996-92-1

IUPAC Name: trimethoxy(phenyl)silane
Synonyms: phenyltrimethoxysilane
Molecular Weight (g/mol): 198.29
Molecular Formula: C9H14O3Si
InChi Key: ZNOCGWVLWPVKAO-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)