missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 85%

Product Code. 10731714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10731714 1 L Glass bottle 1 L
10542072 50 mL Glass bottle 50mL
10603583 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10731714 Supplier Thermo Scientific Acros Supplier No. 370640010
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2996-92-1
Molecular Formula C9H14O3Si
Molecular Weight (g/mol) 198.29
MDL Number MFCD00025689
InChI Key ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Synonym phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
PubChem CID 18137
IUPAC Name trimethoxy(phenyl)silane
SMILES CO[Si](OC)(OC)C1=CC=CC=C1

Spezifikation

Melting Point -25°C
Color Colorless
Density 1.0620g/mL
Boiling Point 233°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula C6H5Si(OCH3)3
Refractive Index 1.4730 to 1.475
Quantity 1 L
Beilstein 16, IV, 1556
Specific Gravity 1.062
Solubility Information Solubility in water: soluble
Formula Weight 198.29
Percent Purity 85%
Physical Form Liquid
Chemical Name or Material Phenyltrimethoxysilane
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Phenyltrimethoxysilane
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.