missing translation for 'onlineSavingsMsg'
Learn More

Phenyltrimethoxysilane, 97%

Product Code. 11494914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494914 25 g 25g
11404924 100 g 100g
11414924 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494914 Supplier Thermo Scientific Alfa Aesar Supplier No. A13827.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2996-92-1 | C9H14O3Si | 198.29 g/mol

Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethoxysilane is used for nanoelectronics and material science research. It is used as crosslinking agent for silicone resin and to produce polymer organic silicon compound.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2996-92-1
Molecular Formula C9H14O3Si
Molecular Weight (g/mol) 198.29
MDL Number MFCD00025689
InChI Key ZNOCGWVLWPVKAO-UHFFFAOYSA-N
Synonym phenyltrimethoxysilane, trimethoxy phenyl silane, silane, trimethoxyphenyl, silane, phenyltrimethoxy, unii-21tqe746s9, trimethoxysilyl benzene, benzene, trimethoxysilyl, phenyl trimethoxysilane, phenyl trimethoxy silane, phenyltri methoxy silane
PubChem CID 18137
IUPAC Name trimethoxy(phenyl)silane
SMILES CO[Si](OC)(OC)C1=CC=CC=C1

Spezifikation

Boiling Point 210°C to 212°C
Melting Point -25°C
Quantity 25 g
Flash Point 86°C (186°F)
Refractive Index 1.4735
Sensitivity Moisture sensitive
Density 1.07
Beilstein 2937896
Solubility Information Reacts with water.
Formula Weight 198.3
Percent Purity 97%
Chemical Name or Material Phenyltrimethoxysilane
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.