accessibility menu, dialog, popup

UserName

N(R)N(S)O(S)(R)NNOHNH2HOOPHOOOH

CAS RN 29984-33-6

IUPAC Name: {[(2R,3S,4S,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: Ara-AMP phosphate, vidarabine monophosphate, adenine arabinoside vidarabine phosphate arabinosyladenine monophosphate monophosphate, vidarabine vidarabine monophosphate ara amp monophosphate, arabinofuranosyladenine vidarabine-5'-monophosphate
Molecular Weight (g/mol): 347.22
Molecular Formula: C10H14N5O7P
InChi Key: UDMBCSSLTHHNCD-UHTZMRCNSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@@H]3O)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results