accessibility menu, dialog, popup

UserName

CAS RN 300-57-2

CH2

CAS RN 300-57-2

IUPAC Name: (prop-2-en-1-yl)benzene
Synonyms: allylbenzene
Molecular Weight (g/mol): 118.18
Molecular Formula: C9H10
InChi Key: HJWLCRVIBGQPNF-UHFFFAOYSA-N
SMILES: C=CCC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)