missing translation for 'onlineSavingsMsg'
Learn More

Allylbenzene, 98%

Product Code. 11429823
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429823 100 g 100g
11419823 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429823 Supplier Thermo Scientific Alfa Aesar Supplier No. A12965.22
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 300-57-2 | C9H10 | 118.179 g/mol

Allylbenzene is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylbenzene is used in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-57-2
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00008651
InChI Key HJWLCRVIBGQPNF-UHFFFAOYSA-N
Synonym allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
PubChem CID 9309
IUPAC Name prop-2-enylbenzene
SMILES C=CCC1=CC=CC=C1

Specifications

Melting Point ∼-50°C
Density 0.892
Boiling Point 158°C to 160°C
Flash Point 33°C (91°F)
Odor Aromatic
Refractive Index 1.51
Quantity 100 g
UN Number UN3295
Beilstein 1098501
Solubility Information Insoluble in water. Soluble in alcohol,ether and benzene.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material Allylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.