missing translation for 'onlineSavingsMsg'
Learn More

Allylbenzene, 98%

Product Code. 11934451 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934451 500 mL Glass bottle 500mL
10785931 25 mL Glass bottle 25mL
10667432 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11934451 Supplier Thermo Scientific Acros Supplier No. 102895000
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 300-57-2 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-57-2
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
InChI Key HJWLCRVIBGQPNF-UHFFFAOYSA-N
Synonym allylbenzene, 2-propenylbenzene, 3-phenyl-1-propene, benzene, 2-propenyl, benzene, allyl, 1-phenyl-2-propene, 3-phenylpropene, 1-propene, 3-phenyl, allyl benzene, 1-benzylethene
PubChem CID 9309
IUPAC Name prop-2-enylbenzene
SMILES C=CCC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 156.0°C to 157.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH2CH=CH2
Refractive Index 1.5100 to 1.5120
Quantity 500 mL
Beilstein 05,484
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble
Formula Weight 118.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allylbenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allylbenzene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.