accessibility menu, dialog, popup

UserName

CAS RN 30084-90-3

IUPAC Name: 9H-fluorene-2-carbaldehyde
Synonyms: 9H-fluorene-2-carbaldehyde
Molecular Weight (g/mol): 194.23
Molecular Formula: C14H10O
InChi Key: MNQGEQSXFDKAPY-UHFFFAOYSA-N
SMILES: O=CC1=CC=C2C(CC3=CC=CC=C23)=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)