missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorenecarboxaldehyde, 99%, ACROS Organics™

Product Code. 10231283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10231283 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10231283 Supplier Thermo Scientific Acros Supplier No. 165500010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 30084-90-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 82°C to 86°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C14H10O
MDL Number MFCD00001123
Quantity 1 g
Beilstein 07, II, 422
Synonym 2-fluorenecarboxaldehyde, fluorene-2-carboxaldehyde, fluorene-2-carbaldehyde, 9h-fluorene-2-carboxaldehyde, 2-formylfluorene, 2-fluorencarbaldehyd, 2-fluorenecarboaldehyde, acmc-1cjjq, 4-07-00-01656 beilstein handbook reference, #
InChI Key MNQGEQSXFDKAPY-UHFFFAOYSA-N
SMILES O=CC1=CC=C2C(CC3=CC=CC=C23)=C1
IUPAC Name 9H-fluorene-2-carbaldehyde
Molecular Weight (g/mol) 194.23
PubChem CID 34804
Formula Weight 194.23
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material 2-Fluorenecarboxaldehyde, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.