CAS RN 30299-08-2
CAS RN 30299-08-2
IUPAC Name:
2-(4-{1-[4-(1-carboxy-1-methylpropoxy)phenyl]cyclohexyl}phenoxy)-2-methylbutanoic acid
Synonyms:
clinofibrate (R*,S*)-clinofibrate (r*,s*)-clinofibrate clinofibrate, (r*,s*)- (R*,S*)-isomer clinofibrate clinofibrate, (r*,s*)-isomer clinofibrato (r*,s*)-isomer clinofibrate 克利贝特 clinofibratum
Molecular Weight (g/mol):
468.59
Molecular Formula:
C28H36O6
InChi Key:
BMOVQUBVGICXQN-UHFFFAOYNA-N
SMILES:
CCC(C)(OC1=CC=C(C=C1)C1(CCCCC1)C1=CC=C(OC(C)(CC)C(O)=O)C=C1)C(O)=O
1
–
1
of
1
results
1
Clinofibrate, MedChemExpress
MedChemExpress Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.