missing translation for 'onlineSavingsMsg'
Learn More

Clinofibrate, MedChemExpress

Product Code. 16461086
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16461086 200 mg 200mg
16446216 100 mg 100mg
16438417 5 mg 5mg
16421897 50 mg 50mg
16405738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16461086 Supplier Medchem Express Supplier No. HY13528
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.

Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.

Specifications

Chemical Name or Material Clinofibrate
CAS 30299-08-2
Color White
Purity Grade Notes Research
Molecular Formula C28H36O6
Quantity 200 mg
Synonym S-8527
Solubility Information DMSO : ≥ 30 mg/mL (64.02 mM)
SMILES CCC(OC1=CC=C(C2(C3=CC=C(OC(CC)(C)C(O)=O)C=C3)CCCCC2)C=C1)(C)C(O)=O
Molecular Weight (g/mol) 468.58
Formula Weight 468.58
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.