CAS RN 3034-04-6
IUPAC Name:
6-methoxy-2-phenyl-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms:
6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one
Molecular Weight (g/mol):
254.29
Molecular Formula:
C16H14O3
InChi Key:
YURQMHCZHLMHIB-UHFFFAOYNA-N
SMILES:
COC1=CC=C2OC(CC(=O)C2=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
6-Methoxyflavanone, 98%, Thermo Scientific™
CAS: 3034-04-6 MDL Number: MFCD00017484