missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxyflavanone, 98%, Thermo Scientific™

Product Code. 11440768
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440768 25 g 25g
11420768 1 g 1g
11430768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440768 Supplier Thermo Scientific Alfa Aesar Supplier No. B25240.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-04-6
MDL Number MFCD00017484

Specifications

Melting Point 141°C to 143°C
Molecular Formula C16H14O3
Quantity 25 g
Beilstein 248912
Synonym 6-methoxyflavanone, 6-methoxy-2-phenylchroman-4-one, 6-methoxy-2-phenyl-chroman-4-one, 6-methoxy-2-phenyl-2,3-dihydro-1-benzopyran-4-one, flavanone, 6-methoxy, acmc-1adv8, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one, 6-methoxy-2-phenyl-2,3-dihydro-4h-chromen-4-one #, 2,3-dihydro-6-methoxy-2-phenyl-4h-1-benzopyran-4-one, 4h-1-benzopyran-4-one, 2,3-dihydro-6-methoxy-2-phenyl
InChI Key YURQMHCZHLMHIB-UHFFFAOYNA-N
SMILES COC1=CC2=C(C=C1)OC(CC2=O)C3=CC=CC=C3
IUPAC Name 6-methoxy-2-phenyl-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 254.29
PubChem CID 97860
Formula Weight 254.29
Percent Purity 98%
Chemical Name or Material 6-Methoxyflavanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.