accessibility menu, dialog, popup

UserName

CAS RN 3061-36-7

OO

CAS RN 3061-36-7

IUPAC Name: 1,4-diphenoxybenzene
Synonyms: 1,4-diphenoxybenzene 4-phenoxydiphenyl oxide hydroquinone diphenyl ether
Molecular Weight (g/mol): 262.31
Molecular Formula: C18H14O2
InChi Key: UVGPELGZPWDPFP-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results