missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diphenoxybenzene, 97%

Product Code. 11482647
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482647 25 g 25g
11472647 5 g 5g
11404220 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482647 Supplier Thermo Scientific Alfa Aesar Supplier No. A14864.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3061-36-7 | C18H14O2 | 262.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3061-36-7
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.308
MDL Number MFCD00038368
InChI Key UVGPELGZPWDPFP-UHFFFAOYSA-N
Synonym benzene, 1,4-diphenoxy, p-diphenoxybenzene, hydroquinone diphenyl ether, p-phenoxyphenoxybenzene, benzene, p-diphenoxy, 4-phenoxydiphenyl oxide, 1,1'-1,4-phenylenebis oxy dibenzene, 1,4-diphenoxy-benzene, acmc-1cqa0
PubChem CID 520487
ChEBI CHEBI:39271
IUPAC Name 1,4-diphenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)OC3=CC=CC=C3

Specifications

Melting Point 72°C to 73°C
Density 1.083
Boiling Point 172°C (0.01 mmHg)
Flash Point 202°C (395°F)
Quantity 25 g
Beilstein 2215945
Formula Weight 262.31
Percent Purity 98%
Chemical Name or Material 1,4-Diphenoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.