accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFFFFFF

CAS RN 307-34-6

IUPAC Name: octadecafluorooctane
Synonyms: perfluorooctane n-perfluorooctane
Molecular Weight (g/mol): 438.06
Molecular Formula: C8F18
InChi Key: YVBBRRALBYAZBM-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results