missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-n-octane, 98%, Thermo Scientific Chemicals

Product Code. 11473338
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11473338

Brand: Thermo Scientific Alfa Aesar A17741.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorooctane may be used in chemical synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-34-6
Molecular Formula C8F18
Molecular Weight (g/mol) 438.059
MDL Number MFCD00042083
InChI Key YVBBRRALBYAZBM-UHFFFAOYSA-N
Synonym perfluorooctane, octadecafluorooctane, n-perfluorooctane, octane, octadecafluoro, eftop ef-l 100, unii-6p60zbk0ql, n-perfluoro octane, perfluoro-n-octane, octadecafluoro-n-octane, 6p60zbk0ql
PubChem CID 9387
ChEBI CHEBI:38826
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-octadecafluorooctane
SMILES C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.73
Boiling Point 99°C to 100°C
Refractive Index 1.282
Quantity 25 g
Beilstein 1717142
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 438.06
Percent Purity 98%
Chemical Name or Material Perfluoro-n-octane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.