accessibility menu, dialog, popup

UserName

FNHSOO

CAS RN 312-63-0

IUPAC Name: N-(4-fluorophenyl)benzenesulfonamide
Synonyms: N-(4-fluorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 251.28
Molecular Formula: C12H10FNO2S
InChi Key: AZORQGXJAXEIGC-UHFFFAOYSA-N
SMILES: FC1=CC=C(NS(=O)(=O)C2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (5)