missing translation for 'onlineSavingsMsg'
Learn More

ELN484228, MedChemExpress

Product Code. 16493256
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493256 25 mg 25mg
16487597 5 mg 5mg
16484848 10 mg 10mg
16475636 100 mg 100mg
16461107 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493256 Supplier Medchem Express Supplier No. HY115038
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress ELN484228 is a blocker of α-synuclein which is a key protein in Parkinson’s disease.

ELN484228 is a blocker of α-synuclein which is a key protein in Parkinson’s disease.

Specifications

Chemical Name or Material ELN484228
CAS 312-63-0
Color White
Purity Grade Notes Research
Molecular Formula C12H10FNO2S
Quantity 25 mg
Solubility Information DMSO : ≥ 50 mg/mL (198.98 mM)
SMILES O=S(C1=CC=CC=C1)(NC2=CC=C(F)C=C2)=O
Molecular Weight (g/mol) 251.28
Formula Weight 251.28
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.