accessibility menu, dialog, popup

UserName

CAS RN 312-94-7

FFFClO

CAS RN 312-94-7

IUPAC Name: 2-(trifluoromethyl)benzoyl chloride
Synonyms: 2-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 208.56
Molecular Formula: C8H4ClF3O
InChi Key: MXIUWSYTQJLIKE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)