missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoyl chloride, 98%, Thermo Scientific™

Product Code. 11433214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433214 100 g 100g
11413214 5 g 5g
11423214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433214 Supplier Thermo Scientific Alfa Aesar Supplier No. A10564.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-94-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000667
InChI Key MXIUWSYTQJLIKE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
PubChem CID 67561
IUPAC Name 2-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=CC=C1C(Cl)=O

Specifications

Melting Point -22°C
Density 1.414
Boiling Point 89°C to 90°C (16 mmHg)
Flash Point 95°C (203°F)
Odor Pungent
Refractive Index 1.479
Quantity 100 g
UN Number UN3265
Beilstein 881040
Sensitivity Moisture sensitive
Formula Weight 208.57
Percent Purity 98%
Chemical Name or Material 2- (Trifluoromethyl)benzoyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.