accessibility menu, dialog, popup

UserName

CAS RN 313-67-7

IUPAC Name: 8-methoxy-6-nitro-2H-phenanthro[3,4-d][1,3]dioxole-5-carboxylic acid
Synonyms: birthwort aristolochin 马兜铃酸 aristolochic acid aristolochic acid I aristolochic acid A aristolochic acid-I
Molecular Weight (g/mol): 341.28
Molecular Formula: C17H11NO7
InChi Key: BBFQZRXNYIEMAW-UHFFFAOYSA-N
SMILES: COC1=C2C=C(C3=C(C=C4OCOC4=C3C2=CC=C1)C(O)=O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)