accessibility menu, dialog, popup

UserName

CH3H3CCH3HSHO

CAS RN 31539-84-1

IUPAC Name: 4-methyl-4-sulfanylpentan-2-ol
Synonyms: 4-methyl-4-sulfanylpentan-2-ol
Molecular Weight (g/mol): 134.24
Molecular Formula: C6H14OS
InChi Key: FDBQLLMYSACLPB-UHFFFAOYNA-N
SMILES: CC(O)CC(C)(C)S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)