accessibility menu, dialog, popup

UserName

OHOCl

CAS RN 31719-77-4

IUPAC Name: 3-(chloromethyl)benzoic acid
Synonyms: 3-(chloromethyl)benzoic acid
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: PBSUMBYSVFTMNG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(CCl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results